2-Phenylethylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Phenethylboronic acid

BB-3483-5G BB-3483-1G BB-3483-25G
COBBBB-3483-5GEA 88.9 EUR
COBBBB-3483-5G COBBBB-3483-1G COBBBB-3483-25G
2-Phenylethylboronsäure
2-Phenylethylboronic acid
Formel: C₈H₁₁BO₂
Molecular Weight: 149.98 g/mol
MDL: MFCD01631226
CAS-Nummer: 34420-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR