2-Phenylethylboronsäure

Lieferant: FLUOROCHEM

Synonyme: Phenethylboronic acid

016267-1G 016267-5G
FLUO016267-1GEA 51.8 EUR
FLUO016267-1G FLUO016267-5G
2-Phenylethylboronsäure
2-Phenylethylboronic acid
Formel: C₈H₁₁BO₂
Molecular Weight: 149.98 g/mol
MDL: MFCD01631226
CAS-Nummer: 34420-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR