(4-Brom-2-nitrophenyl)essigsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Bromo-2-nitrophenylacetic acid
CA-4074-5G
CA-4074-1G
COBBCA-4074-5GEA
274
EUR
COBBCA-4074-5G
COBBCA-4074-1G
(4-Brom-2-nitrophenyl)essigsäure
(4-Brom-2-nitrophenyl)essigsäure
Formel:
C₈H₆BrNO₄ Molecular Weight: 260,0400 g/mol |
MDL:
MFCD05663950 CAS-Nummer: 6127-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...