Trimethylsilyl-2,2-difluor-2-(fluorsulfonyl)acetat
Lieferant: FLUOROCHEM
005917-5G
005917-25G
FLUO005917-5GEA
115
EUR
FLUO005917-5G
FLUO005917-25G
Trimethylsilyl-2,2-difluor-2-(fluorsulfonyl)acetat
Trimethylsilyl-2,2-difluor-2-(fluorsulfonyl)acetat
Formel:
C₅H₉F₃O₄SSi Molecular Weight: 250.27 g/mol |
MDL:
MFCD02093343 CAS-Nummer: 120801-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...