Methyl-2,2-difluor-2-(fluorsulfonyl)acetat ≥97%

Lieferant: Thermo Scientific
Gefahr

H52745.06 H52745.14
H52745.06EA 59.5 EUR
H52745.06 H52745.14
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat ≥97%
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat

Purity: 97% CAS No.: 680-15-9 Molecular Formula: C3H3F3O4S Formula Weight: 192.11 Flash Point: 47°(117°F) Refractive Index: 1.351 Boiling Point: 117-118° Beilstein Reference: 1812896 UN No.: 2920 MDL No.: MFCD00144316

Formel: C₃H₃F₃O₄S
Molecular Weight: 192,11 g/mol
Siedepunkt: 117…118 °C
Dichte: 1,509
Flammpunkt: 47 °C (117 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00144316
CAS-Nummer: 680-15-9
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR