Methyl-2,2-difluor-2-(fluorsulfonyl)acetat
Lieferant: FLUOROCHEM
006805-5G
006805-25G
FLUO006805-5GEA
55.4
EUR
FLUO006805-5G
FLUO006805-25G
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat
Formel:
C₃H₃F₃O₄S Molecular Weight: 192.11 g/mol |
MDL:
MFCD00144316 CAS-Nummer: 680-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...