2,2-Difluor-2-(fluorsulfonyl)essigsäure

Lieferant: FLUOROCHEM

008805-25G 008805-5G
FLUO008805-25GEA 124 EUR
FLUO008805-25G FLUO008805-5G
2,2-Difluor-2-(fluorsulfonyl)essigsäure
2,2-Difluor-2-(fluorsulfonyl)essigsäure
Formel: C₂HF₃O₄S
MDL: MFCD01320807
CAS-Nummer: 1717-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR