2-(4-(Methylsulfonyl)-2-nitrophenyl)essigsäure

Lieferant: COMBI-BLOCKS

CA-4083-5G CA-4083-1G
COBBCA-4083-5GEA 0 EUR
COBBCA-4083-5G COBBCA-4083-1G
2-(4-(Methylsulfonyl)-2-nitrophenyl)essigsäure
2-(4-(Methylsulfonyl)-2-nitrophenyl)essigsäure
Formel: C₉H₉NO₆S
MDL: MFCD09258635
CAS-Nummer: 929000-10-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR