4-(2-Cyanethylcarbamoyl)phenylboronsäurepropandiolcyclischester

Lieferant: COMBI-BLOCKS

PR-3016-5G PR-3016-1G
COBBPR-3016-5GEA 451 EUR
COBBPR-3016-5G COBBPR-3016-1G
4-(2-Cyanethylcarbamoyl)phenylboronsäurepropandiolcyclischester
4-(2-Cyanethylcarbamoyl)phenylboronsäurepropandiolcyclischester
Formel: C₁₃H₁₅BN₂O₃
MDL: MFCD11053850
CAS-Nummer: 1073372-08-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR