3-Acetamido-4-carboxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-6039-1G PN-6039-250M
COBBPN-6039-1GEA 0 EUR
COBBPN-6039-1G COBBPN-6039-250M
3-Acetamido-4-carboxyphenylboronsäurepinakolester
3-Acetamido-4-carboxyphenylboronsäurepinakolester
Formel: C₁₅H₂₀BNO₅
MDL: MFCD13195757
CAS-Nummer: 1218789-96-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR