3-(Dimethylcarbamoyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N,N-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

PN-3050-1G PN-3050-5G
COBBPN-3050-1GEA 137 EUR
COBBPN-3050-1G COBBPN-3050-5G
3-(Dimethylcarbamoyl)phenylboronsäurepinakolester
3-(Dimethylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₂BNO₃
Molecular Weight: 275,1500 g/mol
MDL: MFCD05863907
CAS-Nummer: 832114-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR