4-Carbamoylphenylboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
4-Aminocarbonylphenylboronic acid, pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide, 4-(Aminocarbonyl)phenylboronic acid pinacol ester
021631-5G
021631-1G
FLUO021631-5GEA
434
EUR
FLUO021631-5G
FLUO021631-1G
4-Carbamoylphenylboronsäurepinakolester
4-Carbamoylphenylboronsäurepinakolester
Formel:
C₁₃H₁₈BNO₃ Molecular Weight: 247,0979 g/mol |
MDL:
MFCD05663887 CAS-Nummer: 179117-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...