3-(2-Hydroxyethylcarbamoyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-(2-Hydroxyethyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

PN-8664-1G PN-8664-5G
COBBPN-8664-1GEA 393 EUR
COBBPN-8664-1G COBBPN-8664-5G
3-(2-Hydroxyethylcarbamoyl)phenylboronsäurepinakolester
3-(2-Hydroxyethylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₂BNO₄
Molecular Weight: 291,1500 g/mol
MDL: MFCD06795647
CAS-Nummer: 943911-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR