2-Chlor-5-(methoxycarbonyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Methoxy-5-Methoxycarbonylphenylboronic acid pinacol ester, Methyl 4-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
PN-5015-1G
PN-5015-5G
COBBPN-5015-1GEA
0
EUR
COBBPN-5015-1G
COBBPN-5015-5G
2-Chlor-5-(methoxycarbonyl)phenylboronsäurepinakolester
2-Chlor-5-(methoxycarbonyl)phenylboronsäurepinakolester
Formel:
C₁₅H₂₁BO₅ Molecular Weight: 292.1400 g/mol |
MDL:
MFCD06797995 CAS-Nummer: 269410-10-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...