Ethyl-3-(3-boronobenzamido)propionat

Lieferant: COMBI-BLOCKS

Synonyme: 3-[(3-Ethoxy-3-oxopropyl)carbamoyl]benzeneboronic acid, (3-((3-Ethoxy-3-oxopropyl)carbamoyl)phenyl)boronic acid

BB-3070-5G BB-3070-1G
COBBBB-3070-5GEA 0 EUR
COBBBB-3070-5G COBBBB-3070-1G
Ethyl-3-(3-boronobenzamido)propionat
Ethyl-3-(3-boronobenzamido)propionat
Formel: C₁₂H₁₆BNO₅
MDL: MFCD04115701
CAS-Nummer: 850567-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR