Methyl-3-(3-boronobenzamido)propionat

Lieferant: COMBI-BLOCKS

Synonyme: 3-[(3-Methoxy-3-oxopropyl)carbamoyl]benzeneboronic acid

BB-3420-1G BB-3420-5G
COBBBB-3420-1GEA 0 EUR
COBBBB-3420-1G COBBBB-3420-5G
Methyl-3-(3-boronobenzamido)propionat
Methyl-3-(3-boronobenzamido)propionat
Formel: C₁₁H₁₄BNO₅
MDL: MFCD09475827
CAS-Nummer: 957034-72-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR