Ethyl-3-(4-boronobenzamido)propionat

Lieferant: COMBI-BLOCKS

Synonyme: (4-((3-Ethoxy-3-oxopropyl)carbamoyl)phenyl)boronic acid, 4-[(3-Ethoxy-3-oxopropyl)carbamoyl]benzeneboronic acid

BB-3020-5G BB-3020-1G
COBBBB-3020-5GEA 0 EUR
COBBBB-3020-5G COBBBB-3020-1G
Ethyl-3-(4-boronobenzamido)propionat
Ethyl-3-(4-boronobenzamido)propionat
Formel: C₁₂H₁₆BNO₅
MDL: MFCD04115673
CAS-Nummer: 850568-19-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR