Methyl-3-(4-boronobenzamido)propionat
Lieferant: COMBI-BLOCKS
Synonyme:
4-[(3-Methoxy-3-oxopropyl)carbamoyl]benzeneboronic acid, (4-((3-Methoxy-3-oxopropyl)carbamoyl)phenyl)boronic acid
BB-3419-1G
BB-3419-5G
COBBBB-3419-1GEA
0
EUR
COBBBB-3419-1G
COBBBB-3419-5G
Methyl-3-(4-boronobenzamido)propionat
Methyl-3-(4-boronobenzamido)propionat
Formel:
C₁₁H₁₄BNO₅ Molecular Weight: 251.0400 g/mol |
MDL:
MFCD09475830 CAS-Nummer: 957034-76-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...