3-Acetamido-5-carboxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Acetamido-5-boronobenzoic acid

BB-3479-1G BB-3479-5G
COBBBB-3479-1GEA 420 EUR
COBBBB-3479-1G COBBBB-3479-5G
3-Acetamido-5-carboxyphenylboronsäure
3-Acetamido-5-carboxyphenylboronsäure
Formel: C₉H₁₀BNO₅
Molecular Weight: 222,990 g/mol
MDL: MFCD08436066
CAS-Nummer: 108749-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR