3-(Ethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(N-Ethylaminocarbonyl)phenylboronic acid, 3-(Ethylcarbamoyl)benzeneboronic acid

BB-3057-5G BB-3057-1G
COBBBB-3057-5GEA 912 EUR
COBBBB-3057-5G COBBBB-3057-1G
3-(Ethylcarbamoyl)phenylboronsäure
3-(Ethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₂BNO₃
Molecular Weight: 193,01 g/mol
MDL: MFCD04115689
CAS-Nummer: 850567-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR