3-(2-Methoxyethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3079-1G BB-3079-5G
COBBBB-3079-1GEA 303 EUR
COBBBB-3079-1G COBBBB-3079-5G
3-(2-Methoxyethylcarbamoyl)phenylboronsäure
3-(2-Methoxyethylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₄BNO₄
MDL: MFCD04115710
CAS-Nummer: 850567-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR