3-(Cyclopropylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Cyclopropylaminocarbonyl)phenylboronic acid

BB-3063-5G BB-3063-1G
COBBBB-3063-5GEA 353 EUR
COBBBB-3063-5G COBBBB-3063-1G
3-(Cyclopropylcarbamoyl)phenylboronsäure
3-(Cyclopropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₂BNO₃
MDL: MFCD04115694
CAS-Nummer: 850567-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR