4-(2-Pyridylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Pyridin-2-yl)aminocarbonylphenylboronic acid, (4-(Pyridin-2-ylcarbamoyl)phenyl)boronic acid, 4-(2-Pyridylcarbamoyl)benzeneboronic acid

BB-3032-5G BB-3032-1G
COBBBB-3032-5GEA 2380 EUR
COBBBB-3032-5G COBBBB-3032-1G
4-(2-Pyridylcarbamoyl)phenylboronsäure
4-(2-Pyridylcarbamoyl)phenylboronsäure
Formel: C₁₂H₁₁BN₂O₃
Molecular Weight: 242.04 g/mol
MDL: MFCD06659876
CAS-Nummer: 850568-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR