3-Carboxy-2-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Borono-2-methoxybenzoic acid, 3-Carboxy-2-methoxybenzeneboronic acid

BB-3584-5G BB-3584-1G
COBBBB-3584-5GEA 1590 EUR
COBBBB-3584-5G COBBBB-3584-1G
3-Carboxy-2-methoxyphenylboronsäure
3-Carboxy-2-methoxyphenylboronsäure
Formel: C₈H₉BO₅
Molecular Weight: 195.97 g/mol
MDL: MFCD09027237
CAS-Nummer: 913836-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR