2-Carboxy-3-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-6462-1G
BB-6462-250M
COBBBB-6462-1GEA
0
EUR
COBBBB-6462-1G
COBBBB-6462-250M
2-Carboxy-3-methoxyphenylboronsäure
2-Carboxy-3-methoxyphenylboronsäure
Formel:
C₈H₉BO₅ |
MDL:
MFCD16618937 CAS-Nummer: 1256346-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...