3-Carboxy-4-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Borono-2-methoxybenzoic acid, 3-Carboxy-4-methoxybenzeneboronic acid

BB-3588-5G BB-3588-1G
COBBBB-3588-5GEA 249 EUR
COBBBB-3588-5G COBBBB-3588-1G
3-Carboxy-4-methoxyphenylboronsäure
3-Carboxy-4-methoxyphenylboronsäure
Formel: C₈H₉BO₅
Molecular Weight: 195.97 g/mol
MDL: MFCD06203322
CAS-Nummer: 913836-12-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR