5-Carboxy-2-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Borono-4-methoxybenzoic acid

BB-3593-1G BB-3593-5G
COBBBB-3593-1GEA 126 EUR
COBBBB-3593-1G COBBBB-3593-5G
5-Carboxy-2-methoxyphenylboronsäure
5-Carboxy-2-methoxyphenylboronsäure
Formel: C₈H₉BO₅
Molecular Weight: 195,9700 g/mol
MDL: MFCD06203261
CAS-Nummer: 730971-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR