3-(2-Bromphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3172-1G BB-3172-5G
COBBBB-3172-1GEA 98.3 EUR
COBBBB-3172-1G COBBBB-3172-5G
3-(2-Bromphenylcarbamoyl)phenylboronsäure
3-(2-Bromphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BBrNO₃
MDL: MFCD09027206
CAS-Nummer: 874288-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR