4-(2-Bromphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-((2-Bromophenyl)carbamoyl)phenyl)boronic acid

BB-3122-5G BB-3122-1G
COBBBB-3122-5GEA 547 EUR
COBBBB-3122-5G COBBBB-3122-1G
4-(2-Bromphenylcarbamoyl)phenylboronsäure
4-(2-Bromphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BBrNO₃
Molecular Weight: 319,9500 g/mol
MDL: MFCD09027202
CAS-Nummer: 874288-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR