4-(4-Bromphenylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-((4-Bromophenyl)carbamoyl)phenyl)boronic acid, 4-(4-Bromophenylcarbamoyl)benzeneboronic acid
BB-3120-1G
BB-3120-5G
COBBBB-3120-1GEA
37.4
EUR
COBBBB-3120-1G
COBBBB-3120-5G
4-(4-Bromphenylcarbamoyl)phenylboronsäure
4-(4-Bromphenylcarbamoyl)phenylboronsäure
Formel:
C₁₃H₁₁BBrNO₃ Molecular Weight: 319,95 g/mol |
MDL:
MFCD09027201 CAS-Nummer: 874287-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...