3-Brom-(5-ethoxycarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Bromo-5-(ethoxycarbonyl)phenylboronic acid
BB-3515-5G
BB-3515-1G
COBBBB-3515-5GEA
595
EUR
COBBBB-3515-5G
COBBBB-3515-1G
3-Brom-(5-ethoxycarbonyl)phenylboronsäure
3-Brom-(5-ethoxycarbonyl)phenylboronsäure
Formel:
C₉H₁₀BBrO₄ |
MDL:
MFCD08689516 CAS-Nummer: 913835-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...