3-Brom-(5-methoxycarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Bromo-5-(methoxycarbonyl)phenylboronic acid, (3-Bromo-5-(methoxycarbonyl)phenyl)boronic acid
BB-3514-5G
BB-3514-1G
COBBBB-3514-5GEA
884
EUR
COBBBB-3514-5G
COBBBB-3514-1G
3-Brom-(5-methoxycarbonyl)phenylboronsäure
3-Brom-(5-methoxycarbonyl)phenylboronsäure
Formel:
C₈H₈BBrO₄ Molecular Weight: 258,8617 g/mol |
MDL:
MFCD08689544 CAS-Nummer: 913835-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...