5-Amino-2-brom-4-chlorbenzonitril

Lieferant: COMBI-BLOCKS

AN-2550-5G AN-2550-1G
COBBAN-2550-5GEA 303 EUR
COBBAN-2550-5G COBBAN-2550-1G
5-Amino-2-brom-4-chlorbenzonitril
5-Amino-2-brom-4-chlorbenzonitril
Formel: C₇H₄BrClN₂
MDL: MFCD13195640
CAS-Nummer: 1215206-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR