5-Brom-2-chlor-N,N-dimethylbenzamid

Lieferant: COMBI-BLOCKS

CA-4737-5G CA-4737-25G CA-4737-1G
COBBCA-4737-5GEA 243 EUR
COBBCA-4737-5G COBBCA-4737-25G COBBCA-4737-1G
5-Brom-2-chlor-N,N-dimethylbenzamid
5-Brom-2-chlor-N,N-dimethylbenzamid
Formel: C₉H₉BrClNO
MDL: MFCD04219929
CAS-Nummer: 701258-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR