4-Brom-2-chlor-N,N-dimethylbenzamid

Lieferant: COMBI-BLOCKS

CA-4736-1G CA-4736-5G
COBBCA-4736-1GEA 210 EUR
COBBCA-4736-1G COBBCA-4736-5G
4-Brom-2-chlor-N,N-dimethylbenzamid
4-Brom-2-chlor-N,N-dimethylbenzamid
Formel: C₉H₉BrClNO
MDL: MFCD12756427
CAS-Nummer: 893420-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR