5-Brom-N-tert-butyl-2-chlorbenzamid
Lieferant: COMBI-BLOCKS
CA-4721-1G
CA-4721-5G
COBBCA-4721-1GEA
124
EUR
COBBCA-4721-1G
COBBCA-4721-5G
5-Brom-N-tert-butyl-2-chlorbenzamid
5-Brom-N-tert-butyl-2-chlorbenzamid
Formel:
C₁₁H₁₃BrClNO |
MDL:
MFCD06093685 CAS-Nummer: 892018-58-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...