5-Brom-2-chlor-N-propylbenzamid

Lieferant: COMBI-BLOCKS

CA-4715-5G CA-4715-1G
COBBCA-4715-5GEA 303 EUR
COBBCA-4715-5G COBBCA-4715-1G
5-Brom-2-chlor-N-propylbenzamid
5-Brom-2-chlor-N-propylbenzamid
Formel: C₁₀H₁₁BrClNO
MDL: MFCD05853707
CAS-Nummer: 892018-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR