Methyl-5-brom-4-chlor-2-methoxybenzoat
Lieferant: COMBI-BLOCKS
CA-4475-5G
CA-4475-1G
COBBCA-4475-5GEA
514
EUR
COBBCA-4475-5G
COBBCA-4475-1G
Methyl-5-brom-4-chlor-2-methoxybenzoat
Methyl-5-brom-4-chlor-2-methoxybenzoat
Formel:
C₉H₈BrClO₃ |
MDL:
MFCD09800923 CAS-Nummer: 951885-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...