Methyl-5-brom-4-chlor-2-fluorbenzoat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 5-bromo-4-chloro-2-fluorobenzoate
CA-4429-5G
CA-4429-1G
COBBCA-4429-5GEA
490
EUR
COBBCA-4429-5G
COBBCA-4429-1G
Methyl-5-brom-4-chlor-2-fluorbenzoat
Methyl-5-brom-4-chlor-2-fluorbenzoat
Formel:
C₈H₅BrClFO₂ Molecular Weight: 267.4795 g/mol |
MDL:
MFCD09800902 CAS-Nummer: 951884-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...