4-Brom-N-(3-chlorpropyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0097-5G OT-0097-1G
COBBOT-0097-5GEA 0 EUR
COBBOT-0097-5G COBBOT-0097-1G
4-Brom-N-(3-chlorpropyl)benzolsulfonamid
4-Brom-N-(3-chlorpropyl)benzolsulfonamid
Formel: C₉H₁₁BrClNO₂S
MDL: MFCD01215119
CAS-Nummer: 98768-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR