4-Brom-N,N-diisopropyl-3-methoxybenzamid

Lieferant: COMBI-BLOCKS

CA-4648-5G CA-4648-25G CA-4648-1G
COBBCA-4648-5GEA 303 EUR
COBBCA-4648-5G COBBCA-4648-25G COBBCA-4648-1G
4-Brom-N,N-diisopropyl-3-methoxybenzamid
4-Brom-N,N-diisopropyl-3-methoxybenzamid
Formel: C₁₄H₂₀BrNO₂
MDL: MFCD11504872
CAS-Nummer: 1072944-37-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR