2-Brom-N-cyclohexyl-N-methylbenzamid

Lieferant: COMBI-BLOCKS

CS-9418-5G CS-9418-1G
COBBCS-9418-5GEA 375 EUR
COBBCS-9418-5G COBBCS-9418-1G
2-Brom-N-cyclohexyl-N-methylbenzamid
2-Brom-N-cyclohexyl-N-methylbenzamid
Formel: C₁₄H₁₈BrNO
MDL: MFCD01211895
CAS-Nummer: 349395-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR