2-Brom-N-sec-butylbenzamid

Lieferant: COMBI-BLOCKS

CS-9417-1G CS-9417-5G
COBBCS-9417-1GEA 98.3 EUR
COBBCS-9417-1G COBBCS-9417-5G
2-Brom-N-sec-butylbenzamid
2-Brom-N-sec-butylbenzamid
Formel: C₁₁H₁₄BrNO
MDL: MFCD01212208
CAS-Nummer: 349092-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR