2-Brom-N-tert-butylbenzamid

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-N-(tert-butyl)benzamide

CA-4751-5G CA-4751-1G
COBBCA-4751-5GEA 204 EUR
COBBCA-4751-5G COBBCA-4751-1G
2-Brom-N-tert-butylbenzamid
2-Brom-N-tert-butylbenzamid
Formel: C₁₁H₁₄BrNO
Molecular Weight: 256.1390 g/mol
MDL: MFCD00462994
CAS-Nummer: 168265-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR