2-Brom-N-methylbenzamid
Lieferant: COMBI-BLOCKS
CS-9406-5G
CS-9406-1G
COBBCS-9406-5GEA
432
EUR
COBBCS-9406-5G
COBBCS-9406-1G
2-Brom-N-methylbenzamid
2-Brom-N-methylbenzamid
Formel:
C₈H₈BrNO Molecular Weight: 214,06 g/mol |
MDL:
MFCD00099250 CAS-Nummer: 61436-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...