2-Brom-N-propylbenzamid
Lieferant: COMBI-BLOCKS
CS-9408-5G
CS-9408-1G
COBBCS-9408-5GEA
303
EUR
COBBCS-9408-5G
COBBCS-9408-1G
2-Brom-N-propylbenzamid
2-Brom-N-propylbenzamid
Formel:
C₁₀H₁₂BrNO |
MDL:
MFCD01212206 CAS-Nummer: 346695-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...