3-Brom-N-tert-butylbenzamid
Lieferant: COMBI-BLOCKS
CA-4391-5G
CA-4391-1G
COBBCA-4391-5GEA
595
EUR
COBBCA-4391-5G
COBBCA-4391-1G
3-Brom-N-tert-butylbenzamid
3-Brom-N-tert-butylbenzamid
Formel:
C₁₁H₁₄BrNO |
MDL:
MFCD00751498 CAS-Nummer: 42498-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...