3-Brom-N-butylbenzamid

Lieferant: COMBI-BLOCKS

CA-4494-1G CA-4494-5G
COBBCA-4494-1GEA 98.3 EUR
COBBCA-4494-1G COBBCA-4494-5G
3-Brom-N-butylbenzamid
3-Brom-N-butylbenzamid
Formel: C₁₁H₁₄BrNO
MDL: MFCD00554079
CAS-Nummer: 35390-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR