3-Brom-N-methylbenzamid
Lieferant: COMBI-BLOCKS
CA-4392-5G
CA-4392-1G
COBBCA-4392-5GEA
514
EUR
COBBCA-4392-5G
COBBCA-4392-1G
3-Brom-N-methylbenzamid
3-Brom-N-methylbenzamid
Formel:
C₈H₈BrNO Molecular Weight: 214,0592 g/mol |
MDL:
MFCD01060879 CAS-Nummer: 49834-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...