3-Brom-N-propylbenzamid

Lieferant: COMBI-BLOCKS

CA-4487-5G CA-4487-1G
COBBCA-4487-5GEA 149 EUR
COBBCA-4487-5G COBBCA-4487-1G
3-Brom-N-propylbenzamid
3-Brom-N-propylbenzamid
Formel: C₁₀H₁₂BrNO
MDL: MFCD01215160
CAS-Nummer: 35306-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR